Assay Details
| Gene Symbol: pip5k1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006898.1 | NP_001006899.1 | 1826 | 61 | ||
| GenBank mRNA | BC074662.1 | - | 1826 | 61 | ||
| CT030324.1 | - | 1752 | 61 | |||
More Information
Set Membership: |
|

pip5k1a
Genomic Map