Assay Details
| Gene Symbol: mbd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006900.1 | NP_001006901.1 | 4 - 5 | 515 | 73 | |
| XM_012960212.2 | XP_012815666.1 | 4 - 5 | 725 | 73 | ||
| XM_012960215.2 | XP_012815669.1 | 3 - 4 | 680 | 73 | ||
| XM_018092426.1 | XP_017947915.1 | 4 - 5 | 687 | 73 | ||
| XM_018092431.1 | XP_017947920.1 | 4 - 5 | 684 | 73 | ||
| XM_018092433.1 | XP_017947922.1 | 3 - 4 | 626 | 73 | ||
| GenBank mRNA | BC074695.1 | - | 4 - 5 | 515 | 73 | |
| CR926179.2 | - | 4 - 5 | 636 | 73 | ||
More Information
Set Membership: |
|

mbd3
Genomic Map