Assay Details
| Gene Symbol: agr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006908.1 | NP_001006909.1 | 2 - 3 | 226 | 66 | |
| XM_012964546.2 | XP_012820000.1 | 2 - 3 | 237 | 66 | ||
| GenBank mRNA | BC075325.1 | - | 2 - 3 | 226 | 66 | |
More Information
Set Membership: |
|

agr3
Genomic Map