Assay Details
| Gene Symbol: mal | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006928.2 | NP_001006929.1 | 1 - 2 | 215 | 77 | |
| GenBank mRNA | BC077032.1 | - | 1 - 2 | 219 | 77 | |
| CR760177.2 | - | 1 - 2 | 185 | 77 | ||
More Information
Set Membership: |
|

mal
Genomic Map