Assay Details
| Gene Symbol: ptges3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007104.1 | NP_001007105.1 | 5 - 6 | 477 | 90 | |
| GenBank mRNA | BC084900.1 | - | 5 - 6 | 477 | 90 | |
| CR855476.2 | - | 5 - 6 | 460 | 90 | ||
| CT030427.1 | - | 5 - 6 | 395 | 90 | ||
More Information
Set Membership: |
|

ptges3
Genomic Map