Assay Details
| Gene Symbol: smad9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007478.1 | NP_001007479.1 | 5 - 6 | 1194 | 57 | |
| GenBank mRNA | BC075389.1 | - | 5 - 6 | 1194 | 57 | |
More Information
Set Membership: |
|

smad9
Genomic Map