Assay Details
| Gene Symbol: fancl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007481.1 | NP_001007482.1 | 1 - 2 | 158 | 66 | |
| XM_018093846.1 | XP_017949335.1 | 1 - 2 | 181 | 66 | ||
| XM_018093847.1 | XP_017949336.1 | 1 - 2 | 181 | 66 | ||
| XM_018093848.1 | XP_017949337.1 | 1 - 2 | 181 | 66 | ||
| GenBank mRNA | BC079922.1 | - | 1 - 2 | 158 | 66 | |
| CT027968.1 | - | 1 - 2 | 158 | 66 | ||
More Information
Set Membership: |
|

fancl
Genomic Map