Assay Details
| Gene Symbol: abi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007487.1 | NP_001007488.1 | 9 - 10 | 1345 | 89 | |
| XM_012964547.2 | XP_012820001.1 | 11 - 12 | 1510 | 89 | ||
| XM_012964548.2 | XP_012820002.1 | 10 - 11 | 1491 | 89 | ||
| XM_012964549.2 | XP_012820003.1 | 10 - 11 | 1442 | 89 | ||
| XM_012964550.2 | XP_012820004.1 | 10 - 11 | 1423 | 89 | ||
| XM_012964551.2 | XP_012820005.1 | 9 - 10 | 1355 | 89 | ||
| XM_012964552.2 | XP_012820006.1 | 8 - 9 | 1334 | 89 | ||
| XM_012964553.2 | XP_012820007.1 | 9 - 10 | 1266 | 89 | ||
| XM_012964554.2 | XP_012820008.1 | 8 - 9 | 1180 | 89 | ||
| XM_012964555.2 | XP_012820009.1 | 7 - 8 | 1159 | 89 | ||
| GenBank mRNA | BC079926.1 | - | 9 - 10 | 1345 | 89 | |
More Information
Set Membership: |
|

abi1
Genomic Map