Assay Details
| Gene Symbol: naa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007496.1 | NP_001007497.1 | 5 - 6 | 379 | 124 | |
| XM_012968260.2 | XP_012823714.1 | 5 - 6 | 448 | 124 | ||
| GenBank mRNA | BC079933.1 | - | 5 - 6 | 379 | 124 | |
| CR760841.2 | - | 5 - 6 | 381 | 124 | ||
| CR848272.2 | - | 5 - 6 | 348 | 124 | ||
More Information
Set Membership: |
|

naa10
Genomic Map