Assay Details
| Gene Symbol: bub3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007497.2 | NP_001007498.2 | 7 - 8 | 921 | 83 | |
| GenBank mRNA | BC079934.1 | - | 7 - 8 | 921 | 83 | |
| CR760647.2 | - | 7 - 8 | 918 | 83 | ||
More Information
Set Membership: |
|

bub3
Genomic Map