Assay Details
| Gene Symbol: stac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007506.1 | NP_001007507.1 | 5 - 6 | 647 | 66 | |
| GenBank mRNA | BC079943.1 | - | 5 - 6 | 647 | 66 | |
More Information
Set Membership: |
|

stac3
Genomic Map