Assay Details
| Gene Symbol: naca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007512.1 | NP_001007513.1 | 7 - 8 | 686 | 96 | |
| XM_012956783.2 | XP_012812237.1 | 11 - 12 | 3014 | 96 | ||
| XM_012956784.2 | XP_012812238.1 | 10 - 11 | 2941 | 96 | ||
| XM_012956785.2 | XP_012812239.1 | 12 - 13 | 2728 | 96 | ||
| XM_012956786.2 | XP_012812240.1 | 10 - 11 | 1311 | 96 | ||
| XM_012956787.2 | XP_012812241.1 | 9 - 10 | 1242 | 96 | ||
| XM_012956788.2 | XP_012812242.1 | 9 - 10 | 1155 | 96 | ||
| XM_012956789.1 | XP_012812243.1 | 8 - 9 | 1086 | 96 | ||
| XM_012956790.1 | XP_012812244.1 | 8 - 9 | 903 | 96 | ||
| GenBank mRNA | BC079953.1 | - | 7 - 8 | 686 | 96 | |
| BC167646.1 | - | 8 - 9 | 1953 | 96 | ||
| CT025482.2 | - | 7 - 8 | 651 | 96 | ||
More Information
Set Membership: |
|

naca
Genomic Map