Assay Details
| Gene Symbol: grhpr.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007895.1 | NP_001007896.1 | 6 - 7 | 637 | 95 | |
| GenBank mRNA | BC080331.1 | - | 6 - 7 | 637 | 95 | |
| CR760190.2 | - | 6 - 7 | 634 | 95 | ||
More Information
Set Membership: |
|

grhpr.2
Genomic Map