Assay Details
| Gene Symbol: gdf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007904.1 | NP_001007905.1 | 1 - 2 | 249 | 70 | |
| GenBank mRNA | BC080341.1 | - | 1 - 2 | 249 | 70 | |
| CR760322.2 | - | 1 - 2 | 264 | 70 | ||
More Information
Set Membership: |
|

gdf3
Genomic Map