Assay Details
| Gene Symbol: pla2g12b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007916.1 | NP_001007917.1 | 3 - 4 | 494 | 75 | |
| XM_012966460.2 | XP_012821914.1 | 3 - 4 | 1033 | 75 | ||
| GenBank mRNA | BC080358.1 | - | 3 - 4 | 494 | 75 | |
| CR926443.2 | - | 3 - 4 | 540 | 75 | ||
More Information
Set Membership: |
|

pla2g12b
Genomic Map