Assay Details
| Gene Symbol: rwdd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007934.1 | NP_001007935.1 | 4 - 5 | 517 | 59 | |
| GenBank mRNA | BC080441.1 | - | 4 - 5 | 517 | 59 | |
More Information
Set Membership: |
|

rwdd4
Genomic Map