Assay Details
| Gene Symbol: extl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007951.1 | NP_001007952.1 | 4 - 5 | 563 | 79 | |
| XM_012960912.2 | XP_012816366.1 | 5 - 6 | 79 | |||
| XM_012960913.2 | XP_012816367.1 | 5 - 6 | 79 | |||
| XM_012960914.2 | XP_012816368.1 | 5 - 6 | 79 | |||
| XM_012960915.2 | XP_012816369.1 | 5 - 6 | 79 | |||
| XM_012960917.2 | XP_012816371.1 | 4 - 5 | 79 | |||
| XM_012960918.2 | XP_012816372.1 | 4 - 5 | 79 | |||
| XM_012960919.2 | XP_012816373.1 | 5 - 6 | 79 | |||
| XM_012960920.2 | XP_012816374.1 | 5 - 6 | 79 | |||
| XM_012960922.2 | XP_012816376.1 | 4 - 5 | 79 | |||
| XM_012960923.2 | XP_012816377.1 | 5 - 6 | 79 | |||
| XM_012960924.2 | XP_012816378.1 | 4 - 5 | 79 | |||
| XM_012960925.2 | XP_012816379.1 | 4 - 5 | 79 | |||
| XM_012960926.2 | XP_012816380.1 | 4 - 5 | 79 | |||
| GenBank mRNA | BC080459.1 | - | 4 - 5 | 563 | 79 | |
| CR762109.2 | - | 5 - 6 | 79 | |||
More Information
Set Membership: |
|

extl2
Genomic Map