Assay Details
| Gene Symbol: tfg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007961.2 | NP_001007962.2 | 1 - 2 | 217 | 100 | |
| XM_012956810.2 | XP_012812264.1 | 2 - 3 | 357 | 100 | ||
| GenBank mRNA | BC080473.1 | - | 2 - 3 | 350 | 100 | |
| CT030078.1 | - | 1 - 2 | 217 | 100 | ||
More Information
Set Membership: |
|

tfg
Genomic Map