Assay Details
| Gene Symbol: stmn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007971.1 | NP_001007972.1 | 436 | 76 | ||
| GenBank mRNA | BC080482.1 | - | 436 | 76 | ||
| CT030089.1 | - | 413 | 76 | |||
More Information
Set Membership: |
|

stmn1
Genomic Map