Assay Details
| Gene Symbol: nradd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007998.3 | NP_001007999.2 | 2 - 3 | 491 | 73 | |
| GenBank mRNA | BC080877.1 | - | 2 - 3 | 455 | 73 | |
More Information
Set Membership: |
|

nradd
Genomic Map