Assay Details
| Gene Symbol: clp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008001.1 | NP_001008002.1 | 2 - 3 | 703 | 79 | |
| XM_018095355.1 | XP_017950844.1 | 2 - 3 | 706 | 79 | ||
| GenBank mRNA | BC080880.1 | - | 2 - 3 | 703 | 79 | |
| CR848531.2 | - | 2 - 3 | 712 | 79 | ||
More Information
Set Membership: |
|

clp1
Genomic Map