Assay Details
| Gene Symbol: pdcd6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008003.1 | NP_001008004.1 | 3 - 4 | 230 | 78 | |
| XM_012964564.2 | XP_012820018.1 | 3 - 4 | 656 | 78 | ||
| GenBank mRNA | BC080882.1 | - | 3 - 4 | 230 | 78 | |
| CR760932.2 | - | 3 - 4 | 360 | 78 | ||
More Information
Set Membership: |
|

pdcd6
Genomic Map