Assay Details
| Gene Symbol: f11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008021.1 | NP_001008022.1 | 256 | 64 | ||
| GenBank mRNA | BC080901.1 | - | 256 | 64 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

f11r| Gene Symbol: f11r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008021.1 | NP_001008022.1 | 256 | 64 | ||
| GenBank mRNA | BC080901.1 | - | 256 | 64 | ||
Set Membership: |
|