Assay Details
Gene Symbol: keap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008023.1 | NP_001008024.1 | 1641 | 75 | ||
GenBank mRNA | BC080903.1 | - | 1641 | 75 | ||
CR855517.2 | - | 1635 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
