Assay Details
| Gene Symbol: ears2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008041.1 | NP_001008042.1 | 7 - 8 | 1359 | 67 | |
| GenBank mRNA | BC080925.1 | - | 7 - 8 | 1359 | 67 | |
More Information
Set Membership: |
|

ears2
Genomic Map