Assay Details
| Gene Symbol: cptp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008042.1 | NP_001008043.1 | 2 - 3 | 184 | 66 | |
| XM_012966470.2 | XP_012821924.1 | 2 - 3 | 289 | 66 | ||
| XM_012966471.1 | XP_012821925.1 | 2 - 3 | 285 | 66 | ||
| XM_012966472.2 | XP_012821926.1 | 2 - 3 | 879 | 66 | ||
| XM_012966473.1 | XP_012821927.1 | 2 - 3 | 288 | 66 | ||
| GenBank mRNA | BC080926.1 | - | 2 - 3 | 184 | 66 | |
More Information
Set Membership: |
|

cptp
Genomic Map