Assay Details
| Gene Symbol: fst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008056.1 | NP_001008057.1 | 4 - 5 | 812 | 80 | |
| XM_012960359.2 | XP_012815813.1 | 4 - 5 | 1247 | 80 | ||
| XM_012960363.2 | XP_012815817.1 | 4 - 5 | 1247 | 80 | ||
| GenBank mRNA | BC080943.1 | - | 4 - 5 | 812 | 80 | |
More Information
Set Membership: |
|

fst
Genomic Map