Assay Details
| Gene Symbol: rflnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008058.1 | NP_001008059.1 | 2 - 3 | 431 | 109 | |
| GenBank mRNA | BC080945.1 | - | 2 - 3 | 431 | 109 | |
More Information
Set Membership: |
|

rflnb
Genomic Map