Assay Details
| Gene Symbol: impdh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008065.1 | NP_001008066.1 | 5 - 6 | 592 | 59 | |
| XM_012960932.2 | XP_012816386.1 | 5 - 6 | 615 | 59 | ||
| GenBank mRNA | BC080955.1 | - | 5 - 6 | 592 | 59 | |
| CR760493.2 | - | 5 - 6 | 579 | 59 | ||
More Information
Set Membership: |
|

impdh2
Genomic Map