Assay Details
| Gene Symbol: rab5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008067.1 | NP_001008068.1 | 5 - 6 | 739 | 92 | |
| GenBank mRNA | BC080959.1 | - | 5 - 6 | 739 | 92 | |
| CR855837.1 | - | 5 - 6 | 670 | 92 | ||
More Information
Set Membership: |
|

rab5a
Genomic Map