Assay Details
| Gene Symbol: serhl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008070.1 | NP_001008071.1 | 7 - 8 | 502 | 94 | |
| XM_012960935.2 | XP_012816389.1 | 6 - 7 | 934 | 94 | ||
| XM_018093019.1 | XP_017948508.1 | 7 - 8 | 954 | 94 | ||
| GenBank mRNA | BC080964.1 | - | 7 - 8 | 502 | 94 | |
More Information
Set Membership: |
|

serhl
Genomic Map