Assay Details
| Gene Symbol: alas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008071.1 | NP_001008072.1 | 5 - 6 | 857 | 62 | |
| GenBank mRNA | BC080965.1 | - | 5 - 6 | 857 | 62 | |
| CR760758.2 | - | 5 - 6 | 849 | 62 | ||
More Information
Set Membership: |
|

alas1
Genomic Map