Assay Details
| Gene Symbol: natd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008089.1 | NP_001008090.1 | 4 - 5 | 542 | 67 | |
| GenBank mRNA | BC081288.1 | - | 4 - 5 | 542 | 67 | |
| CR848524.2 | - | 4 - 5 | 515 | 67 | ||
More Information
Set Membership: |
|

natd1
Genomic Map