Assay Details
| Gene Symbol: nck1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008100.1 | NP_001008101.1 | 4 - 5 | 1128 | 68 | |
| XM_012970334.2 | XP_012825788.1 | 5 - 6 | 1263 | 68 | ||
| XM_012970335.2 | XP_012825789.1 | 5 - 6 | 1516 | 68 | ||
| XM_012970336.2 | XP_012825790.1 | 2 - 3 | 826 | 68 | ||
| XM_018093859.1 | XP_017949348.1 | 4 - 5 | 1221 | 68 | ||
| GenBank mRNA | BC081302.1 | - | 4 - 5 | 1128 | 68 | |
More Information
Set Membership: |
|

nck1
Genomic Map