Assay Details
| Gene Symbol: cidec | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008101.1 | NP_001008102.1 | 3 - 4 | 245 | 67 | |
| XM_012960954.2 | XP_012816408.1 | 2 - 3 | 789 | 67 | ||
| XM_012960955.2 | XP_012816409.1 | 4 - 5 | 544 | 67 | ||
| XM_012960956.2 | XP_012816410.1 | 2 - 3 | 233 | 67 | ||
| XM_012960957.2 | XP_012816411.1 | 2 - 3 | 218 | 67 | ||
| XM_018093023.1 | XP_017948512.1 | 3 - 4 | 442 | 67 | ||
| GenBank mRNA | BC081304.1 | - | 3 - 4 | 245 | 67 | |
| CR761715.2 | - | 3 - 4 | 253 | 67 | ||
More Information
Set Membership: |
|

cidec
Genomic Map