Assay Details
| Gene Symbol: rab33b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008108.1 | NP_001008109.1 | 2 - 3 | 522 | 80 | |
| XM_012960439.2 | XP_012815893.1 | 2 - 3 | 613 | 80 | ||
| GenBank mRNA | BC081311.1 | - | 2 - 3 | 522 | 80 | |
| CR760567.2 | - | 2 - 3 | 563 | 80 | ||
More Information
Set Membership: |
|

rab33b
Genomic Map