Assay Details
| Gene Symbol: pin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008109.1 | NP_001008110.1 | 3 - 4 | 418 | 81 | |
| XM_012953001.2 | XP_012808455.1 | 3 - 4 | 392 | 81 | ||
| GenBank mRNA | BC081312.1 | - | 3 - 4 | 418 | 81 | |
| CR760780.2 | - | 3 - 4 | 373 | 81 | ||
More Information
Set Membership: |
|

pin1
Genomic Map