Assay Details
| Gene Symbol: dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008111.1 | NP_001008112.1 | 2 - 3 | 296 | 92 | |
| GenBank mRNA | BC081315.1 | - | 2 - 3 | 296 | 92 | |
| CR762160.2 | - | 2 - 3 | 273 | 92 | ||
More Information
Set Membership: |
|

dnajb1
Genomic Map