Assay Details
| Gene Symbol: dnajb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008111.1 | NP_001008112.1 | 3 - 4 | 903 | 89 | |
| GenBank mRNA | BC081315.1 | - | 3 - 4 | 903 | 89 | |
More Information
Set Membership: |
|

dnajb1
Genomic Map