Assay Details
| Gene Symbol: pim1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008131.1 | NP_001008132.1 | 2 - 3 | 340 | 70 | |
| XM_012960961.2 | XP_012816415.1 | 2 - 3 | 342 | 70 | ||
| XM_012960962.2 | XP_012816416.1 | 2 - 3 | 485 | 70 | ||
| XM_018093025.1 | XP_017948514.1 | 2 - 3 | 1165 | 70 | ||
| GenBank mRNA | BC081340.1 | - | 2 - 3 | 340 | 70 | |
| CR760428.2 | - | 2 - 3 | 279 | 70 | ||
More Information
Set Membership: |
|

pim1
Genomic Map