Assay Details
| Gene Symbol: cdk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008135.1 | NP_001008136.1 | 6 - 7 | 866 | 66 | |
| GenBank mRNA | BC081346.1 | - | 6 - 7 | 866 | 66 | |
More Information
Set Membership: |
|

cdk2
Genomic Map