Assay Details
| Gene Symbol: tbxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008138.1 | NP_001008139.1 | 5 - 6 | 847 | 78 | |
| GenBank mRNA | BC081350.1 | - | 5 - 6 | 847 | 78 | |
More Information
Set Membership: |
|

tbxt
Genomic Map