Assay Details
| Gene Symbol: tbxt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008138.1 | NP_001008139.1 | 6 - 7 | 1026 | 77 | |
| XM_012962922.1 | XP_012818376.1 | 6 - 7 | 1109 | 77 | ||
| GenBank mRNA | BC081350.1 | - | 6 - 7 | 1026 | 77 | |
| CR761440.2 | - | 6 - 7 | 1060 | 77 | ||
More Information
Set Membership: |
|

tbxt
Genomic Map