Assay Details
| Gene Symbol: strada | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008150.1 | NP_001008151.1 | 11 - 12 | 1194 | 67 | |
| XM_012971690.2 | XP_012827144.1 | 12 - 13 | 1283 | 67 | ||
| XM_012971691.2 | XP_012827145.1 | 11 - 12 | 1254 | 67 | ||
| XM_012971692.2 | XP_012827146.1 | 10 - 11 | 1192 | 67 | ||
| XM_012971693.2 | XP_012827147.1 | 9 - 10 | 1134 | 67 | ||
| XM_018097717.1 | XP_017953206.1 | 11 - 12 | 1221 | 67 | ||
| GenBank mRNA | BC081364.1 | - | 11 - 12 | 1194 | 67 | |
More Information
Set Membership: |
|

strada
Genomic Map