Assay Details
| Gene Symbol: mpp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008151.2 | NP_001008152.1 | 6 - 7 | 779 | 82 | |
| XM_018093026.1 | XP_017948515.1 | 6 - 7 | 1033 | 82 | ||
| GenBank mRNA | BC081365.1 | - | 6 - 7 | 784 | 82 | |
| CT027847.1 | - | 6 - 7 | 753 | 82 | ||
More Information
Set Membership: |
|

mpp1
Genomic Map