Assay Details
| Gene Symbol: kpna2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008154.2 | NP_001008155.1 | 7 - 8 | 1001 | 60 | |
| XM_012966478.2 | XP_012821932.1 | 6 - 7 | 1063 | 60 | ||
| XM_012966479.2 | XP_012821933.1 | 7 - 8 | 1094 | 60 | ||
| GenBank mRNA | BC081368.1 | - | 7 - 8 | 1003 | 60 | |
| CR762026.2 | - | 6 - 7 | 974 | 60 | ||
More Information
Set Membership: |
|

kpna2
Genomic Map