Assay Details
| Gene Symbol: cbs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008170.1 | NP_001008171.1 | 1152 | 60 | ||
| GenBank mRNA | BC082489.1 | - | 1152 | 60 | ||
| CR942417.2 | - | 1144 | 60 | |||
More Information
Set Membership: |
|

cbs
Genomic Map