Assay Details
| Gene Symbol: cyct | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008175.1 | NP_001008176.1 | 2 - 3 | 258 | 84 | |
| XM_012970350.2 | XP_012825804.1 | 3 - 4 | 502 | 84 | ||
| GenBank mRNA | BC082495.1 | - | 2 - 3 | 258 | 84 | |
| CR760771.2 | - | 1 - 2 | 195 | 84 | ||
More Information
Set Membership: |
|

cyct
Genomic Map