Assay Details
| Gene Symbol: utp14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008204.1 | NP_001008205.1 | 6 - 7 | 623 | 104 | |
| XM_012968277.2 | XP_012823731.1 | 6 - 7 | 599 | 104 | ||
| XM_012968278.2 | XP_012823732.1 | 6 - 7 | 680 | 104 | ||
| GenBank mRNA | BC080963.1 | - | 6 - 7 | 623 | 104 | |
More Information
Set Membership: |
|

utp14a
Genomic Map