Assay Details
| Gene Symbol: bap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001008205.1 | NP_001008206.1 | 2 - 3 | 233 | 82 | |
| GenBank mRNA | BC080985.1 | - | 2 - 3 | 233 | 82 | |
| CR848566.2 | - | 2 - 3 | 129 | 82 | ||
More Information
Set Membership: |
|

bap1
Genomic Map